O-Benzylhydroxylamine hydrochloride ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B1079-25G
B1079-250G
B1079-5G
TCIAB1079-25GEA
52.8
EUR
TCIAB1079-25G
TCIAB1079-250G
TCIAB1079-5G
O-Benzylhydroxylamine hydrochloride ≥98.0% (by titrimetric analysis)
O-Benzylhydroxylamine hydrochloride
Formule:
C₇H₁₀ClNO |
MDL Nummer:
MFCD00012952 CAS nummer: 2687-43-6 EINECS: 220-249-0 |
|
Specification Test Results
Appearance | White to Almost white powder to crystaline |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...