Ethyl-2-oxocyclohexanecarboxylate ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Ethyl 2-oxocyclohexanecarboxylate
C0913-25ML
C0913-500ML
C0913-100ML
TCIAC0913-25MLEA
47.3
EUR
TCIAC0913-25ML
TCIAC0913-500ML
TCIAC0913-100ML
Ethyl-2-oxocyclohexanecarboxylate ≥95.0% (by GC)
Ethyl-2-oxocyclohexanecarboxylate
Formule:
C₉H₁₄O₃ MW: 170.21 g/mol Kookpunt: 160°C Vlampunt: 85°C |
MDL Nummer:
MFCD00001631 CAS nummer: 1655-07-8 EINECS: 216-731-5 |
|
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Acid Value | max. 5.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...