4-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
T2481-1G T2481-5G
TCIAT2481-1GEA 28.6 EUR
TCIAT2481-1G TCIAT2481-5G
4-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde ≥97.0% (by GC)
4-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde
Formule: C₉H₆F₄O₂
MW: 222.1364 g/mol
Kookpunt: 103°C
Smeltpunt: 9°C
MDL Nummer: MFCD00039577
CAS nummer: 35295-36-4
EINECS: 252-497-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=995c3136808acfaa4bc5cd759815db5174a68b60&vid=995c3136808acfaa4bc5cd759815db5174a68b60&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %