(Benzoylmethylene)triphenylphosphorane ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

T2002-5G T2002-1G
TCIAT2002-5GEA 154 EUR
TCIAT2002-5G TCIAT2002-1G
(Benzoylmethylene)triphenylphosphorane ≥98.0% (by titrimetric analysis)
(Benzoylmethylene)triphenylphosphorane
Formule: C₂₆H₂₁OP
Smeltpunt: 188°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00014088
CAS nummer: 859-65-4
EINECS: 212-727-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53cbffdc7169a2bd9905f19acb05df5937646eed&vid=53cbffdc7169a2bd9905f19acb05df5937646eed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 184.0 to 188.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR