Coumalic acid ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Oxo-2H-pyran-5-carboxylic acid

C0391-25G C0391-5G
TCIAC0391-25GEA 176 EUR
TCIAC0391-25G TCIAC0391-5G
Coumalic acid ≥97.0% (by GC, titration analysis)
Coumalic acid
Formule: C₆H₄O₄
Kookpunt: 218°C
MDL Nummer: MFCD00006644
CAS nummer: 500-05-0
EINECS: 207-899-0
Merck Index: 14,02557
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=850b1cd7a8ded3d7f5c7a631ec33c078a10ddfb9&vid=850b1cd7a8ded3d7f5c7a631ec33c078a10ddfb9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR