4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P1184-1G
P1184-5G
P1184-25G
TCIAP1184-1GEA
57.2
EUR
TCIAP1184-1G
TCIAP1184-5G
TCIAP1184-25G
4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione
4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione
Formule:
C₈H₅N₃O₂ Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD00003148 CAS nummer: 4233-33-4 EINECS: 224-191-7 |
|
Specification Test Results
Appearance | Red to Dark red to Dark purple powder to crystal |
Elemental analysis (Nitrogen) | 23.50 to 24.30 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...