1-Naphthoyl chloride ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
A14460.09 A14460.18 A14460.30
A14460.09EA 37.1 EUR
A14460.09 A14460.18 A14460.30
1-Naphthoyl chloride ≥98%
1-Naphthoyl chloride
Purity: 98% CAS No.: 879-18-5 Molecular Formula: C11H7ClO Formula Weight: 190.63 Flash Point: 110°(230°F) Refractive Index: 1.6525 Odour: Pungent Melting Point: 16-19° Boiling Point: 118°/0.1mm Beilstein Reference: 775785 UN No.: 3265 MDL No.: MFCD00004002
Formule: C₁₁H₇ClO
Kookpunt: 118 °C (0,1 mmHg)
Smeltpunt: 16…19 °C
Dichtheid: 1,265
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004002
CAS nummer: 879-18-5
EINECS: 212-903-9
UN: 3265
ADR: 8,II