1,1,3,3-Tetrabromoacetone ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1,1,3,3-Tetrabromo-2-propanone
T1026-25G T1026-5G
TCIAT1026-25GEA 303 EUR
TCIAT1026-25G TCIAT1026-5G
1,1,3,3-Tetrabromoacetone ≥98.0% (by GC)
1,1,3,3-Tetrabromoacetone
Formule: C₃H₂Br₄O
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00059470
CAS nummer: 22612-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24c4ce067774407ba1689d4c2520babda4889b60&vid=24c4ce067774407ba1689d4c2520babda4889b60&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C
Solubility in Methanol almost transparency