Trimethyl phosphonoacetate 96,0 GC_ASSAY_METHOD

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Methyl 2-(dimethoxyphosphoryl)acetate, Dimethyl methoxycarbonylmethanephosphonate

P1265-25G P1265-250G
TCIAP1265-25GEA 29.7 EUR
TCIAP1265-25G TCIAP1265-250G
Trimethyl phosphonoacetate 96,0 GC_ASSAY_METHOD
Trimethyl phosphonoacetate
Formule: C₅H₁₁O₅P
MW: 182.11 g/mol
Kookpunt: 108°C
Vlampunt: 70°C
MDL Nummer: MFCD00008452
CAS nummer: 5927-18-4
EINECS: 227-663-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8d53f6bb2a6db730ebcb6beaa430dafc205078d&vid=e8d53f6bb2a6db730ebcb6beaa430dafc205078d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR