5-Bromo-1,3-benzodioxole ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B1230-25G B1230-5G
TCIAB1230-25GEA 63.8 EUR
TCIAB1230-25G TCIAB1230-5G
5-Bromo-1,3-benzodioxole ≥98.0% (by GC)
5-Bromo-1,3-benzodioxole
Formule: C₇H₅BrO₂
Kookpunt: 228°C
MDL Nummer: MFCD00005821
CAS nummer: 2635-13-4
EINECS: 220-123-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=739b27da5fead3813e02db8c0163a58ff9f440f3&vid=739b27da5fead3813e02db8c0163a58ff9f440f3&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) min. 98.0 %