Diethyl (3-methylbenzyl)phosphonate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Diethyl 3-methylbenzylphosphonate

D3328-25G D3328-5G
TCIAD3328-25GEA 273 EUR
TCIAD3328-25G TCIAD3328-5G
Diethyl (3-methylbenzyl)phosphonate ≥98.0% (by GC)
Diethyl (3-methylbenzyl)phosphonate
Formule: C₁₂H₁₉O₃P
MW: 242.2512 g/mol
Kookpunt: 127°C
MDL Nummer: MFCD08276364
CAS nummer: 63909-50-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c994d5798560acfd0c95f19e35299248199a4416&vid=c994d5798560acfd0c95f19e35299248199a4416&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR