Diethyl (2-bromoethyl)phosphonate ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B1781-25G B1781-5G
TCIAB1781-25GEA 219 EUR
TCIAB1781-25G TCIAB1781-5G
Diethyl (2-bromoethyl)phosphonate ≥95.0% (by GC)
Diethyl (2-bromoethyl)phosphonate
Formule: C₆H₁₄BrO₃P
MW: 245.06 g/mol
Kookpunt: 115°C
MDL Nummer: MFCD00000239
CAS nummer: 5324-30-1
EINECS: 226-193-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dd7003559f80af727c080df49cf58c26a193ed6&vid=9dd7003559f80af727c080df49cf58c26a193ed6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %