Diethyl (methoxymethyl)phosphonate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D3873-25G D3873-5G
TCIAD3873-25GEA 263 EUR
TCIAD3873-25G TCIAD3873-5G
Diethyl (methoxymethyl)phosphonate ≥98.0% (by GC)
Diethyl (methoxymethyl)phosphonate
Formule: C₆H₁₅O₄P
Kookpunt: 88°C
CAS nummer: 32806-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d28961aff04e75e1ac796b4a38935321d09c0699&vid=d28961aff04e75e1ac796b4a38935321d09c0699&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR