1,3-Dichloroacetone 50% in 1,3-dichloroacetone
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB01010.1 SB01010.10 SB01010.50
MAYBSB01010.1EA 0 EUR
MAYBSB01010.1 MAYBSB01010.10 MAYBSB01010.50
1,3-Dichloroacetone 50% in 1,3-dichloroacetone
1,3-Dichloroacetone
Formule: ClH₂CC(O)CH₂Cl
MW: 126.97 g/mol
Kookpunt: 173
Vlampunt: 95
MDL Nummer: MFCD00000937
CAS nummer: 534-07-6
EINECS: 208-585-6
Merck Index: 13,03077