3-Bromo-4-fluorophenylacetic acid ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B2976.0005 B2976-25G B2976-5G
TCIAB2976.0005EA 0 EUR
TCIAB2976.0005 TCIAB2976-25G TCIAB2976-5G
3-Bromo-4-fluorophenylacetic acid ≥97.0% (by GC, titration analysis)
3-Bromo-4-fluorophenylacetic acid
Formule: C₈H₆BrFO₂
MW: 233.04 g/mol
MDL Nummer: MFCD04038373
CAS nummer: 194019-11-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11549661883db3793979c907a60495e5b23933a0&vid=11549661883db3793979c907a60495e5b23933a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR