4-Acetamidobenzenesulphonyl azide ≥97%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Purity: 97%
CAS No.: 2158-14-7
Molecular Formula: C8H8N4O3S
Formula Weight: 240.24
Storage Temperature: -30°C to -10°C
Melting Point: 107-111°
Beilstein Reference: 2219568
MDL No.: MFCD00029626
Formule:
C₈H₈N₄O₃S Smeltpunt: 107…111 °C |
MDL Nummer:
MFCD00029626 CAS nummer: 2158-14-7 |