2,4-Dichloro-1-(2-nitrovinyl)benzene
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: trans-2,4-Dichloro-β-nitrostyrene
S01394.1 S01394.10 S01394.50
MAYBS01394.1EA 0 EUR
MAYBS01394.1 MAYBS01394.10 MAYBS01394.50
2,4-Dichloro-1-(2-nitrovinyl)benzene
2,4-Dichloro-1-(2-nitrovinyl)benzene
Formule: C₈H₅Cl₂NO₂
MW: 218.037 g/mol
MDL Nummer: MFCD00053063
CAS nummer: 18984-21-9