Benzyl isocyanide ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B2185-1G B2185-5G
TCIAB2185-1GEA 70.4 EUR
TCIAB2185-1G TCIAB2185-5G
Benzyl isocyanide ≥96.0% (by GC)
Benzyl isocyanide
Formule: C₈H₇N
Kookpunt: 106°C
Vlampunt: 79°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00000004
CAS nummer: 10340-91-7
EINECS: 233-738-9
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=000b3aca719d475b61f64c3ff6f8963a814cc0df&vid=000b3aca719d475b61f64c3ff6f8963a814cc0df&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 96.0 %