Diethyl (4-bromobenzyl)phosphonate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Diethyl 4-bromobenzylphosphonate, (4-Bromobenzyl)phosphonic acid diethyl ester
D3688-25G
D3688-5G
TCIAD3688-25GEA
244
EUR
TCIAD3688-25G
TCIAD3688-5G
Diethyl (4-bromobenzyl)phosphonate ≥98.0% (by GC)
Diethyl (4-bromobenzyl)phosphonate
Formule:
C₁₁H₁₆BrO₃P MW: 307.12 g/mol Kookpunt: 183°C |
MDL Nummer:
MFCD05663693 CAS nummer: 38186-51-5 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...