Diethyl (3-chlorobenzyl)phosphonate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Diethyl 3-chlorobenzylphosphonate, (3-Chlorobenzyl)phosphonic acid diethyl ester

C1595-1G C1595-5G
TCIAC1595-1GEA 49.5 EUR
TCIAC1595-1G TCIAC1595-5G
Diethyl (3-chlorobenzyl)phosphonate ≥98.0% (by GC)
Diethyl (3-chlorobenzyl)phosphonate
Formule: C₁₁H₁₆ClO₃P
MW: 262.6697 g/mol
Kookpunt: 163°C
MDL Nummer: MFCD01861282
CAS nummer: 78055-64-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51eba04e1b8311361c61e116d08561c69ce22eba&vid=51eba04e1b8311361c61e116d08561c69ce22eba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR