Diethyl (4-cyanobenzyl)phosphonate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Diethyl 4-cyanobenzylphosphonate
D3323-25G D3323-5G
TCIAD3323-25GEA 454 EUR
TCIAD3323-25G TCIAD3323-5G
Diethyl (4-cyanobenzyl)phosphonate ≥98.0% (by GC)
Diethyl (4-cyanobenzyl)phosphonate
Formule: C₁₂H₁₆NO₃P
MW: 253.234 g/mol
Kookpunt: 169°C
Smeltpunt: 31°C
MDL Nummer: MFCD08274488
CAS nummer: 1552-41-6
UN: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5c1954a392a2e476ce9c4a23e7651e8c95d3f29&vid=b5c1954a392a2e476ce9c4a23e7651e8c95d3f29&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %