Diethyl (4-aminobenzyl)phosphonate 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B22279.06 B22279.14 B22279.22
B22279.06EA 70.3 EUR
B22279.06 B22279.14 B22279.22
Diethyl (4-aminobenzyl)phosphonate 99%
Diethyl (4-aminobenzyl)phosphonate
Purity: 99% CAS No.: 20074-79-7 Molecular Formula: C11H18NO3P Formula Weight: 243.24 Melting Point: 91-94° Beilstein Reference: 2941613
Formule: C₁₁H₁₈NO₃P
MW: 243.24 g/mol
Smeltpunt: 91…94 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007913
CAS nummer: 20074-79-7
EINECS: 243-504-8