Diethyl (4-methoxybenzyl)phosphonate ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Diethyl 4-methoxybenzylphosphonate
D4000-25G D4000-5G
TCIAD4000-25GEA 194 EUR
TCIAD4000-25G TCIAD4000-5G
Diethyl (4-methoxybenzyl)phosphonate ≥95.0% (by GC)
Diethyl (4-methoxybenzyl)phosphonate
Formule: C₁₂H₁₉O₄P
Kookpunt: 151°C
MDL Nummer: MFCD00015662
CAS nummer: 1145-93-3
EINECS: 214-548-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=764f9d108ead6cc3326023f0c0e67e5fbd6d357c&vid=764f9d108ead6cc3326023f0c0e67e5fbd6d357c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %