Diethyl cyanophosphonate ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Diethyl phosphorocyanidate
C1242-25G
C1242-5G
TCIAC1242-25GEA
218
EUR
TCIAC1242-25G
TCIAC1242-5G
Diethyl cyanophosphonate ≥95.0% (by GC)
Diethyl cyanophosphonate
Formule:
C₅H₁₀NO₃P MW: 163.11 g/mol Kookpunt: 104°C Vlampunt: 81°C |
MDL Nummer:
MFCD00010256 CAS nummer: 2942-58-7 EINECS: 220-936-5 UN: 2922 ADR: 8 III |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...