6-Methoxy-2-naphthoic acid ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M1418-25G M1418-5G
TCIAM1418-25GEA 186 EUR
TCIAM1418-25G TCIAM1418-5G
6-Methoxy-2-naphthoic acid ≥97.0% (by GC, titration analysis)
6-Methoxy-2-naphthoic acid
Formule: C₁₂H₁₀O₃
MW: 202.21 g/mol
MDL Nummer: MFCD00092750
CAS nummer: 2471-70-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63e68d9c1550141f3ebdb5aebcacdebc586339a2&vid=63e68d9c1550141f3ebdb5aebcacdebc586339a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR