Thiophen-2-acetonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T0799-25ML T0799-5ML
TCIAT0799-25MLEA 114 EUR
TCIAT0799-25ML TCIAT0799-5ML
Thiophen-2-acetonitrile ≥98.0% (by GC)
Thiophen-2-acetonitrile
Formule: C₆H₅NS
Kookpunt: 120°C
Vlampunt: 102°C
MDL Nummer: MFCD00005453
CAS nummer: 20893-30-5
EINECS: 244-104-6
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ae354ea89e01a8bb70a9eaa4e3607e6b9e3e1a2&vid=4ae354ea89e01a8bb70a9eaa4e3607e6b9e3e1a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR