2-Acetylbenzo[b]thiophene ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
2-Acetylbenzothiophene
B21173.06
B21173.03
B21173.06EA
160
EUR
B21173.06
B21173.03
2-Acetylbenzo[b]thiophene ≥98%
2-Acetylbenzo[b]thiophene
Purity: 98%
CAS No.: 22720-75-8
Molecular Formula: C10H8OS
Formula Weight: 176.23
Melting Point: 86-88°
MDL No.: MFCD00090217
Formule:
C₁₀H₈OS Smeltpunt: 86…88 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00090217 CAS nummer: 22720-75-8 EINECS: 245-177-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...