Fmoc-Met-Opfp

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4011860.0005 4011860.0025
BACE4011860.0005EA 0 EUR
BACE4011860.0005 BACE4011860.0025
Fmoc-Met-Opfp
Fmoc-Met-Opfp
Formule: C₂₆H₂₀F₅NO₄S
MDL Nummer: MFCD00065665
CAS nummer: 86060-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR