(+)-Dehydroabietylamine ≥90.0% (by GC) optical resolving agent

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D1588-25G D1588-5G
TCIAD1588-25GEA 160 EUR
TCIAD1588-25G TCIAD1588-5G
(+)-Dehydroabietylamine ≥90.0% (by GC) optical resolving agent
(+)-Dehydroabietylamine
Formule: C₂₀H₃₁N
MDL Nummer: MFCD00213430
CAS nummer: 1446-61-3
EINECS: 215-899-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1825e38514ed43f7201947f1304aee560053b9e&vid=661e1508a44665c0c35035cb58bcbdbc9a3e226c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 90.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 38.0 to 42.0 deg-C
Specific rotation [a]20/D +40.0 to +50.0 deg (C=4,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR