(R)-(-)-α-Cyclohexanemethylamine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: (R)-(-)-1-Cyclohexylethylamine, (R)-1-Cyclohexylethylamine

C1541-25G C1541-5G
TCIAC1541-25GEA 389 EUR
TCIAC1541-25G TCIAC1541-5G
(R)-(-)-α-Cyclohexanemethylamine ≥98.0% (by GC, titration analysis)
(R)-(-)-α-Cyclohexanemethylamine
Formule: C₈H₁₇N
MW: 127.23 g/mol
Kookpunt: 60°C
Vlampunt: 51°C
MDL Nummer: MFCD00043338
CAS nummer: 5913-13-3
EINECS: 227-634-2
UN: 2734
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e98b3773f87a7a66a56a1c6823f8948e5b832467&vid=e98b3773f87a7a66a56a1c6823f8948e5b832467&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 93.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR