(S)-(+)-α-Cyclohexanemethylamine ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
(S)-1-Cyclohexylethanamine, (S)-(+)-1-Cyclohexylethylamine
C1531-25G
C1531-5G
TCIAC1531-25GEA
116
EUR
TCIAC1531-25G
TCIAC1531-5G
(S)-(+)-α-Cyclohexanemethylamine ≥98.0% (by GC, titration analysis)
(S)-(+)-α-Cyclohexanemethylamine
Formule:
C₈H₁₇N MW: 127.2273 g/mol Kookpunt: 74°C Vlampunt: 52°C |
MDL Nummer:
MFCD00066261 CAS nummer: 17430-98-7 UN: 2734 ADR: 8 II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Optical purity (GC) | min. 95.0 ee% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...