3H-1,2-Benzodithiol-one-1,1-dioxide (beaucage reagent) ≥99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L20129.03
L20129.06
L20129.MD
L20129.03EA
68.9
EUR
L20129.03
L20129.06
L20129.MD
3H-1,2-Benzodithiol-one-1,1-dioxide (beaucage reagent) ≥99%
3H-1,2-Benzodithiol-one-1,1-dioxide (beaucage reagent)
Purity: 99%
CAS No.: 66304-01-6
Molecular Formula: C7H4O3S2
Formula Weight: 200.24
Melting Point: 102-107°
Beilstein Reference: 4180332
UN No.: 3335
MDL No.: MFCD00132960
Formule:
C₇H₄O₃S₂ Smeltpunt: 102…107 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00132960 CAS nummer: 66304-01-6 ADR: 9,III |