Einhorn's saccharometer
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41370010 41370020
HECH41370010EA 23.4 EUR
HECH41370010 HECH41370020
Einhorn's saccharometer
Hydrometers
Fermentation saccharometers are used for the estimation of sugar in urine and diabetes diagnostic.
The carbon dioxide created in the fermenting process would rise to the top of the closed tube and force the level of liquid down.
The percentage of sugar could be read off the graduated scale on the closed tube side of the apparatus.