Tightening tools for Very High Pressure (VHP) fittings, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHP-2000 VHP-1000
554-2198EA 1380 EUR
554-2198 554-1999
Tightening tools for Very High Pressure (VHP) fittings, Upchurch Scientific®
Moersleutels
  • Configured for optimum torque to ensure a strong connection
  • Prolongs the lifetime of reusable fittings by not over-tightening
  • Available for multiple fitting head styles
Certificaten: Calibrated according to ISO 6789:2003