Microorganisms, EZ-Accu Shot™
Leverancier: MICROBIOLOGICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0443A 0484A
MCBL0443AEA 0 EUR
MCBL0443A MCBL0484A
Microorganisms, EZ-Accu Shot™
Testsystemen voor microbiologie Micro-organisme testen
EZ-Accu Shot provides a fast, convenient and reliable solution for growth promotion testing, media challenge testing and more.
  • Delivers 10 to 100 CFU per 0,1 ml
  • Strains are less than or equal to four passages from reference culture
  • Instant dissolve pellet reduces preparation time
  • No dilutions required for simplified test procedure
  • Up to 8 hours stability allows for ultimate flexibility
  • 50 tests per kit, ideal quantity for labs of any size
  • Refrigerated storage is easy and economical
  • Traceability to reference cultures ensures authenticity
  • Online Certificate of Analysis provides detailed strain information
  • Technical Support experts available for guidance
Each instant-dissolve microorganism pellet is designed to deliver 10 to 100 CFU per inoculum and provides 8 hours of stability after rehydration.
Certificaten: Meets USP/Ph. Eur./JP and Pharmacopeia guidelines.
Leveringsinformatie: EZ-Accu Shot™ kits include: 5 vials of a single enumerated microorganism (1 lyophilised pellet per vial), 5 vials of hydrating fluid (1,2 ml in each vial), Peel-Off certificate of assay.