VWR® 'Trunk' Carrier for Transport of Secondary Containers for Biological Samples
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PBIB39179EA 256 EUR
PBIB39179 PBIB39180 PBIB89932 PBIB91303
VWR® 'Trunk' Carrier for Transport of Secondary Containers for Biological Samples
Dozen Transportdozen
Insulated waterproof carrier.
  • Can be disinfected with water solution or the most common disinfectants
  • Easy carriage: Lightweight and provided with shoulder strap for easy transport of samples
  • Emergency procedures and safety instructions indelibly printed on lid internal holders for eutectic plates in order to allow transportation at refrigerated temperature over short distances
  • Document pocket: For holding sample cards
For transport of secondary containers for biological samples - conforms to World Health Organisation (WHO/EMC/97,3) guidelines. The TRUNK carriers are to be used as: 'External container into which secondary containers are placed'.