Demeter pipettes
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42574001 42574002
HECH42574001EA 8.4 EUR
HECH42574001 HECH42574002
Demeter pipettes
Pipetten Gegradueerde pipetten
Diluting pipettes, soda glass, with ring marks.
  • Fused amber graduation 'ASSISTENT-PERMANENT'
  • Calibrated for blowing out up to 1 ml
  • Other marks are calibrated to delivery ('EX')
Certificaten: DIN 12838, DIN 12839