Accessories for stopwatches
Leverancier: Hanhart
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HANH905.0698-40EA 14.6 EUR
HANH905.0698-40
Accessories for stopwatches
Timers Mechanische timers
Hanhart offers a calibration service which includes calibration of watches and a certificate of their accuracy.
Calibration certificates can be issued for the following watches:
All mechanical stopwatches except for the 'Felix' range.
The electronic stopwatches Delta, Modul, Accord, Profil, Magma, Stopstar, Spectron, Mesotron, Prisma.