Graduated pipettes, with piston
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42548020 42548048 42548052 42548036 42548056 42548044
HECH42548020EA 21.8 EUR
HECH42548020 HECH42548048 HECH42548052 HECH42548036 HECH42548056 HECH42548044
Graduated pipettes, with piston
Pipetten Gegradueerde pipetten
Safety measuring pipettes with interchangeable ground-in piston, standard type.
  • Tolerance 1,5 times higher than the official tolerance of class AS
  • Fused amber graduation 'ASSISTENT-PERMANENT'
  • Zero at top, calibrated
  • Fire-polished tip
Verpakking: Each pipette is packed in a plastic tube.