Swabs tubes
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SWA6_1000 SWA5_1000
GREISWA6_1000EA 91.3 EUR
GREISWA6_1000 GREISWA5_1000
Swabs tubes
Wattenstaafjes Wattenstaafjes voor transport
Swab in tubes suitable for the collection and transport of non-human bacteriological, serological, and cytological samples.
  • Sterilised by irradiation
  • Swab tubes made of polystyrene
Useful for hygienic controls in veterinary research as well as in environmental sampling.
Verpakking: Individually wrapped.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904
Order Now
Specificaties
Environmentally Preferable