HPLC columns, Acclaim™ mixed-mode WCX-1
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
068352 068353 068371 069705 070092 070093 071910 071911
555-7163EA 1180 EUR
555-7163 555-7164 555-7165 555-7137 555-7205 555-7206 555-7168 555-7169
HPLC columns, Acclaim™ mixed-mode WCX-1
Kolommen voor chromatografie
These are high-efficiency, silica-based columns, manufactured by bonding a specially designed proprietary ligand with both hydrophobic and weak cation-exchange properties. Selectivity of ionisable and neutral compounds can be controlled independently or simultaneously by tuning mobile phase ionic strength, pH or organic modifier.
  • Adjustable selectivity
  • Selectivity complementary to RP columns
  • Ideal selectivity for separating basic molecules
  • Multimode separation mechanism: RP, cation-exchange, ion-exclusion and HILIC
  • Capable of separating a mixture of neutral and basic compounds
Particle type: high purity, spherical, porous silica
Max. pressure: 5800 psi (4500 psi for 3,0×50 mm column)
Flow rate range: 0,1 to 0,6 ml/min for 2,1 mm ID formats; 0,2 to 1,2 ml/min for 3,0 mm ID formats; 0,5 to 3,0 ml/min for 4,6 mm ID formats
pH range: 2,5 to 7,0
Max. temperature: 50 °C