Solid phase extraction cartridges, Bond Elut™ Diol
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12256007 12113009 12102036 12102009 12102067 12113035
VARI12256007EA 218 EUR
VARI12256007 VARI12113009 551-0320 551-0305 VARI12102067 VARI12113035
Solid phase extraction cartridges, Bond Elut™ Diol
Kolommen voor chromatografie
Bond Elut™ Diol (2OH) resembles unbonded silica in its tendency for strong hydrogen bonding with analytes. 2OH can also be employed in the non-polar mode because the hydrocarbon spacer on its functional group provides enough non-polar character for retention of hydrophobic analytes. Bond Elut™ Diol is a listed SPE device for the DIN 14333-1 method on benzimidazole fungicides.
  • Provides polar and non-polar modes
  • Strong hydrogen bonding with analytes
  • Resembles unbonded silica in its capabilities