Solid phase extraction cartridges, Bond Elut™ ENV
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12105014 12105015 12255011 12255012 12105012 12255014 12105013 12105016
551-0335EA 185 EUR
551-0335 551-0336 551-0402 551-0403 551-0334 VARI12255014 VARI12105013 VARI12105016
Solid phase extraction cartridges, Bond Elut™ ENV
Kolommen voor chromatografie
Bond Elut™ ENV, a PS/DVB polymer, is designed for the extraction of polar organic residues. It contains 125 μm spherical particles with a high degree of cross linking, advantageous for high volume, fast flow-through applications.
  • Provides polar and non-polar modes
  • Strong hydrogen bonding with analytes
  • Resembles unbonded silica in its capabilities