Solid phase extraction cartridges, Bond Elut™ NEXUS and NEXUS WCX
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12113100 12103101 12113101 12103100 12253102 12253101 12103102 12102157
551-0352EA 177 EUR
551-0352 551-0330 551-0353 VARI12103100 VARI12253102 VARI12253101 VARI12103102 VARI12102157
Solid phase extraction cartridges, Bond Elut™ NEXUS and NEXUS WCX
Kolommen voor chromatografie
Bond Elut™ NEXUS is an ultra-clean polymeric sorbent which has bi-modal porosity and a high surface area. NEXUS offers a nonpolar retention mechanism with no pre-conditioning required. The large particle size makes NEXUS ideal for extractions from highly viscous samples, such as horse urine.
Based on the same base polymer technology, Bond Elut™ NEXUS WCX is a weak cation exchange sorbent that offers extra selectivity for analytes, such as quaternary ammonium drugs and anabolic steroids.
  • Large particle size allows excellent flow for viscous samples
  • Non-conditioning method saves time and
More Product Information
Order Now
Specificaties
Environmentally Preferable
Oppervlakte 575 m²/g 575 m²/g 575 m²/g