Dichloro(phenyl)phosphine ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: P,P-Dichlorophenylphosphine, Phenyldichlorophosphine
P0207-25ML P0207-500ML
TCIAP0207-25MLEA 17.6 EUR
TCIAP0207-25ML TCIAP0207-500ML
Dichloro(phenyl)phosphine ≥98.0% (by titrimetric analysis)
Dichloro(phenyl)phosphine
Formule: C₆H₅Cl₂P
MW: 178.98 g/mol
Kookpunt: 59°C
Smeltpunt: -51
Vlampunt: 102°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000528
CAS nummer: 644-97-3
EINECS: 211-425-8
UN: 2798
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c5cd3518e40314e6cc6bf7449dcdba0ff1830b8&vid=8c5cd3518e40314e6cc6bf7449dcdba0ff1830b8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (Argentmetric Titration) min. 98.0 %