Span® 85

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Arlacel 85, Sorbitan trioleate

S0064-25G S0064-500G
TCIAS0064-25GEA 18.7 EUR
TCIAS0064-25G TCIAS0064-500G
Span® 85
Span® 85
Formule: C₆₀H₁₀₈O₈
Kookpunt: ? 100 °C (1013 hPa)
Smeltpunt: –23°C
Dichtheid: 0,95 g/cm³ (20 °C)
Vlampunt: 253°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00133820
CAS nummer: 26266-58-0
EINECS: 247-569-3

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green clear liquid
Saponification Value 163.0 to 190.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR