Octanoyl chloride ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

O0039-25ML O0039-500ML O0039-100ML
TCIAO0039-25MLEA 22 EUR
TCIAO0039-25ML TCIAO0039-500ML TCIAO0039-100ML
Octanoyl chloride ≥99.0% (by GC)
Octanoyl chloride
Formule: C₈H₁₅ClO
Kookpunt: 196 °C (1013 hPa)
Smeltpunt: –63 °C
Dichtheid: 0,95 g/cm³ (20 °C)
Vlampunt: 85°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000766
CAS nummer: 111-64-8
EINECS: 203-891-6
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba03a51a0a5dd7eb42982f69c1db396de0701063&vid=ba03a51a0a5dd7eb42982f69c1db396de0701063&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Purity (Argentmetric Titration) 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR