2,3-Dimethyl-1,3-butadiene ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Stabilisatie: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
D1148-10ML D1148-25ML
TCIAD1148-10MLEA 44 EUR
TCIAD1148-10ML TCIAD1148-25ML
2,3-Dimethyl-1,3-butadiene ≥98.0% (by GC) stabilised
2,3-Dimethyl-1,3-butadiene
Formule: C₆H₁₀
Kookpunt: 69 °C (1013 hPa)
Smeltpunt: –76 °C
Dichtheid: 0,7261 g/cm³ (20 °C)
Vlampunt: -1°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00008595
CAS nummer: 513-81-5
EINECS: 208-172-0
UN: 3295
ADR: 3,II
Merck Index: 14,03238
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %