Phenyltrimethoxysilane ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
P0857-25ML P0857-500ML P0857-100ML
TCIAP0857-25MLEA 17.6 EUR
TCIAP0857-25ML TCIAP0857-500ML TCIAP0857-100ML
Phenyltrimethoxysilane ≥98.0% (by GC)
Phenyltrimethoxysilane
Formule: C₉H₁₄O₃Si
Kookpunt: 131°C
Dichtheid: 1,064 g/cm³ (20 °C)
Vlampunt: 94°C
MDL Nummer: MFCD00025689
CAS nummer: 2996-92-1
EINECS: 221-066-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9493bb4429e3e01bd6bd74121c6058a8553b387&vid=d9493bb4429e3e01bd6bd74121c6058a8553b387&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %