5-Iodo-m-xylene ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Stabilisatie:
Stabilised with copper
I0548-25G
I0548-5G
TCIAI0548-25GEA
88
EUR
TCIAI0548-25G
TCIAI0548-5G
5-Iodo-m-xylene ≥98.0% (by GC) stabilised
5-Iodo-m-xylene
Formule:
C₈H₉I Kookpunt: 229°C Dichtheid: 1,61 g/cm³ (20 °C) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00060659 CAS nummer: 22445-41-6 |
|
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...